missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies (Z)-Guggulsterone
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
- (Z)-Guggulsterone is a plant steroid found in the resin of the guggul plant Commiphora mukul that acts as a selective antagonist of farnesoid X receptor (FXR; Cui et al.) It decreases chenodeoxycholic acid-induced FXR activation (IC50 = 10 μM) in the presence of 100 μM chenodeoxycholic acid (Cui et al.; Urizar et al.)
- Induces differentiation of human pluripotent stem cell-derived neural stem cells into dopaminergic neurons (Gonzalez et al.)
- Decreases hepatic low-density lipoprotein cholesterol and triglyceride levels in rodents fed with a high-cholesterol diet (Urizar et al.)
- Regulates growth and metastasis of tumor cells in human multiple myeloma cells by the expression of STAT3-regulated antiapoptotic (Bcl-2, Bcl-xL, and Mcl-1), proliferative (cyclin D1), and angiogenic (VEGF) gene products (Ahn et al.)
- Inhibits tube formation in human umbilical vein endothelial cells (HUVECs) and migration in HUVECs or a human prostate cancer cell line (Xiao et al.)
Specifications
Specifications
| CAS | 39025-23-5 |
| Quantity | 50 mg |
| Molecular Formula | C21H28O2 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction