missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 1 Peptide

Product Code. 12010176
Change view
Click to view available options
Quantity:
800 x 20 μL Assays
Unit Size:
800 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12010176 800 x 20 μL Assays 800 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12010176 Supplier Invitrogen™ Supplier No. PV3174
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Z'-LYTE™ Kinase Assay Kit - Ser/Thr 1 Peptide

A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon a porcine pyruvate liver kinase derived peptide used to study PKA substrate specificity, and containing an R-R-X-S consensus sequence, where X is any residue.
TRUSTED_SUSTAINABILITY

Specifications

Target Serine/Threonine Kinases
For Use With (Application) FRET
Assay Serine/Threonine Kinase Assay
Content And Storage Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions.
Assay Entry Biochemical phospho-specific activity (no antibody)
Label or Dye Methylcoumarin
Product Type Kinase Assay Kit
Quantity 800 x 20 μL Assays
Format 384-well plate
Sample Type porcine pyruvate liver kinase (LRRASLG)
Excitation/Emission 405⁄465, 535
No. of Reactions 800 Reactions
References BBRC vol. 61, pp 559-563 (1974), JBC, vol 252, p 4888 (1978)
Readout End Point
Shipping Condition Dry Ice
Substrate Peptide-Based Substrate
Target Entry PRKX, PRKG2, RPS6KA5, PRKACA, PRKG1, RPS6KA4, AURKB, AURKA
Validated Kinases AURKA, AURKB, PRKACA, PRKG1, PRKG2, PRKX, RPS6KA4, RPS6KA5
Product Line Z´-LYTE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.