missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 12 Peptide

Product Code. 12040186
Change view
Click to view available options
Quantity:
800 x 20μL, 800 x 20 μL Assays
Unit Size:
800 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12040186 800 x 20μL, 800 x 20 μL Assays 800 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12040186 Supplier Thermo Scientific™ Supplier No. PV3673
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format.

A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon CDK7, containing Ser-164 and a (S/T)-P-X-(K/R) consensus sequence, where X is any residue.

TRUSTED_SUSTAINABILITY

Specifications

Description Z'-LYTE™ Ser/Thr 12 Peptide Kinase Assay Kit, 800 x 20μL, 800rxns, Serine/Threonine Kinase Assay Type, Peptide-Based Substrate, 535nm Emission, 405/465nm Excitation, Methylcoumarin Label, End Point Readout, Serine/Threonine Kinases Target Enzyme, FRET Technique, Used for Pharma and Biopharma, Protein Assays and Analysis
Product Line Z ́-LYTE
For Use With (Application) Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, z and acutelyte™ kinase assays, enzyme and protein activity assays, protein assays and analysis, kinase activity, FRET
Includes Z'-LYTE™ Ser/Thr 11 peptide, Z'-LYTE™ Ser/Thr 11 phospho-peptide, 5X kinase buffer A, ATP, development reagent B, development buffer
Sample Volume 800 x 20μL
Content And Storage –80°C, Z'-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions.
Assay Serine/Threonine Kinase Assay
Assay Entry Biochemical phospho-specific activity (no antibody)
Product Type Z'-LYTE™ Ser/Thr 12 Peptide Kinase Assay Kit
Quantity 800 x 20μL, 800 x 20 μL Assays
Format 384-well plate
Excitation/Emission 405/465, 535 nm
Label or Dye Methylcoumarin
No. of Reactions 800 Reactions
Readout End Point
References Mol. Cell. Biol. Vol 21. pp. 88-99 (2001)
Shipping Condition Dry Ice
Source Based on CDK7 (GLAKSFGSPNRAY(pT)H)
Substrate Properties Peptide-Based Substrate
Target Entry CDK1, CDK2, CDK5
Target Enzyme Serine/Threonine Kinases
Validated Kinases CDK1, CDK2, CDK5
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.