missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 22 Peptide
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon CDK7, containing Ser-161.
Specifications
Specifications
| Description | Z'-LYTE™ Ser/Thr 22 Peptide Kinase Assay Kit, 800 x 20μL, 800rxns, Serine/Threonine Kinase Assay Type, Peptide-Based Substrate, 535nm Emission, 405/465nm Excitation, Methylcoumarin Label, End Point Readout, Serine/Threonine Kinases Target Enzyme, FRET Technique, Used for Pharma and Biopharma, Protein Assays and Analysis |
| Product Line | Z ́-LYTE |
| For Use With (Application) | Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, z and acute-lyte™ kinase assays, enzyme and protein activity assays, protein assays and, FRET |
| Includes | Z'-LYTE™ Ser/Thr 11 peptide, Z'-LYTE™ Ser/Thr 11 phospho-peptide, 5X kinase buffer A, ATP, development reagent B, development buffer |
| Sample Volume | 800 x 20μL |
| Content And Storage | –80°C, Z'-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. |
| Assay | Serine/Threonine Kinase Assay |
| Assay Entry | Biochemical phospho-specific activity (no antibody) |
| Product Type | Z'-LYTE™ Ser/Thr 22 Peptide Kinase Assay Kit |
| Quantity | 800 x 20μL, 800 x 20 μL Assays |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?