missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 3 Peptide
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon bovine MBP Thr-97 derived peptide, containing a P-X-(S/T)-P consensus sequence, where X is any residue.
Specifications
Specifications
| Description | Z'-LYTE™ Kinase Assay Kit - Ser/Thr 3 Peptide, 800 x 20μL Assays, 800 rxns, 405/465nm Excitation, 535nm Emission, Methylcoumarin Label, FRET Technique, Serine/Threonine Kinases Target Enzyme, Peptide-Based Substrate Properties, End Point Readout, Used For Pharma and Biopharma, Protein Assays and Analysis |
| Product Line | Z ́-LYTE |
| For Use With (Application) | Pharma and biopharma, protein assays and analysis, target and lead identification and validation, FRET |
| Includes | All necessary reagents for performing primary or secondary screens |
| Sample Volume | 800 x 20μL |
| Content And Storage | –80°C, Z'-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. |
| Assay | Serine/Threonine Kinase Assay |
| Assay Entry | Biochemical phospho-specific activity (no antibody) |
| Product Type | Z'-LYTE™ Kinase Assay Kit - Ser/Thr 3 Peptide |
| Quantity | 800 x 20μL, 800 x 20 μL Assays |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?