missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Tyrosine 4 Peptide

Product Code. 12000186
Change view
Click to view available options
Quantity:
800 x 20 μL Assays
Unit Size:
800 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12000186 800 x 20 μL Assays 800 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12000186 Supplier Invitrogen™ Supplier No. PV3193
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Z'-LYTE™ Kinase Assay Kit - Tyrosine 4 Peptide

A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide contains an X-(E⁄D)-Y-(I⁄L⁄V) consensus sequence, where X is any residue.
TRUSTED_SUSTAINABILITY

Specifications

Product Line Z´-LYTE
Target Tyrosine Kinases
For Use With (Application) FRET
Assay Tyrosine Kinase Assay
Content And Storage Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions.
Assay Entry Biochemical phospho-specific activity (no antibody)
Label or Dye Methylcoumarin
Product Type Kinase Assay Kit
Quantity 800 x 20 μL Assays
Format 384-well plate
Excitation/Emission 405⁄465, 535
No. of Reactions 800 Reactions
Readout End Point
Shipping Condition Dry Ice
Research Category Signal Transduction
Substrate Peptide-Based Substrate
Target Entry PDGFRA T674I, PDGFRB, FGFR1, FGFR2, EGFR, FGFR4, FLT1, MUSK, JAK3, FLT4, FGFR3, INSRR, PDGFRA D842V, EPHA2, PDGFRA, PDGFRA V561D
Validated Kinases EGFR, EPHA2, FGFR, FLT1, FLT4, INSRR, JAK3, MUSK, PDGFRA, PDGFRB
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.