missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Nonacos-20-en-12-one, TRC

Product Code. 30483354
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483354 50 mg -
30474017 5 mg -
30472728 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483354 Supplier Toronto Research Chemicals Supplier No. TRCN64977050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1027944-51-9
Molecular Formula C29H56O
Molecular Weight (g/mol) 420.75
IUPAC Name (Z)-nonacos-20-en-12-one
SMILES CCCCCCCCCCCC(=O)CCCCCCC\C=C/CCCCCCCC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C29H56O/c1-3-5-7-9-11-13-14-15-16-17-18-20-22-24-26-28-29(30)27-25-23-21-19-12-10-8-6-4-2/h15-16H,3-14,17-28H2,1-2H3/b16-15-
Formula Weight 420.4331
Chemical Name or Material (Z)-Nonacos-20-en-12-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.