missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Z-VAD(OMe)-Fluromethylketone Inhibitor

Product Code. 11424906 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
3 mg
5 mg
Unit Size:
1mg
3mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424906 5 mg 5mg
11404906 1 mg 1mg
11414906 3 mg 3mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424906 Supplier MP Biomedicals™ Supplier No. 03FK00905
  • ONLINE EXCLUSIVE PRICE
    1580.00 €
    1498.35 € / 5mg
    Save 81.65 €
    5% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Z-VAD(OMe)-FMK inhibitor synthesized using proprietary technology is cell permeable and irreversibly binds to activated Caspases to block apoptosis.

Z-VAD(OMe)-FMK is an irreversible and cell permeable broad-spectrum Caspase Inhibitor. It has been shown to inhibit Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine).

Specifications

Color White
Content And Storage 4°C
Quantity 5 mg
Product Type Z-Val-Ala-Asp-(OMe)-Fluromethylketone Inhibitor
Molecular Weight (g/mol) 467.487
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.