Learn More
Tocris Bioscience™ Z-VRPR-FMK trifluoroacetate salt
Irreversible MALT1 inhibitor
Brand:  Tocris Bioscience™ 4645/500U
Encompass
Product Code. 15766565
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Z-VRPR-FMK trifluoroacetate salt is an irreversible MALT1 inhibitor. Suppresses T cell activation-induced cleavage of Bcl-10 in a dose-dependent manor. Reduces Jurkat cell adhesion to fibronectin. Cell permeable.
This peptide is supplied in gross weight.
Specifications
Specifications
10892
MALT1
500 μg
Store at -20°C
Observed M.W.: 790.81
C31H49FN10O6.CF3CO2H