missing translation for 'onlineSavingsMsg'
Learn More

(Z,Z)-Alpha-Furil Dioxime, TRC

Product Code. 30396849
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396849 10 mg 10mg
30405394 50 mg -
30402101 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396849 Supplier Toronto Research Chemicals Supplier No. TRCF86465310MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23789-34-6
Molecular Formula C10H8N2O4
Molecular Weight (g/mol) 220.18
SMILES O\N=C(\C(=N\O)\c1occc1)/c2occc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H8N2O4/c13-11-9(7-3-1-5-15-7)10(12-14)8-4-2-6-16-8/h1-6,13-14H/b11-9+,12-10+
Chemical Name or Material (Z,Z)-alpha-Furil Dioxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.