Learn More
LNI Swissgas ZA FID AIR 1.5
Product Code. 17842269 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage unitSize
17842269 100/115/220/230 V Each
1 options
Product Code. 17842269 Supplier LNI Swissgas Supplier No. 6920.25.015
Zero air generator for FID.
  • Zero air generator for FID 1.5 L/min.
Specifications
Voltage 100/115/220/230 V
Type Zero Air Gas Generator