Description
- Zero air generator 30,000 cc/min HC/carbon dioxide/moisture removal.
- Needs source of dry and oil free air.
Specifications
Specifications
| Voltage | 100/115/220/230 V |
| Type | Zero Air Gas Generator |