Learn More
LNI Swissgas ZA TOTAL 6000
Product Code. 17832009 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage unitSize
17832009 100/115/220/230 V Each
1 options
Product Code. 17832009 Supplier LNI Swissgas Supplier No. 6920.40.060
  • ONLINE EXCLUSIVE PRICE
    6670.00 €
    5784.46 € / Pack of 1
    Save 885.54 €
    13% Off
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Zero air generator for HC/carbon dioxide/moisture removal.
  • Zero air generator 6000 cc/min HC/carbon dioxide/moisture removal.
  • Needs source of dry and oil free air.
Specifications
Voltage 100/115/220/230 V
Type Zero Air Gas Generator