Learn More
LNI Swissgas ZA TOTAL C 1.5
Product Code. 17832099 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage Unit Size
17832099 100/115/220/230 V Each
1 options
Product Code. 17832099 Supplier LNI Swissgas Supplier No. 6920.25.215
  • ONLINE EXCLUSIVE PRICE
    8400.00 €
    7284.13 € / Pack of 1
    Save 1115.87 €
    13% Off
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Zero air generator for HC/carbon dioxide/moisture removal with integrated compressor.
  • Zero air generator 1.5 L/min HC/carbon dioxide/moisture removal with integrated compressor.
Specifications
Voltage 100/115/220/230 V
Type Zero Air Gas Generator