Learn More
LNI Swissgas ZA TOTAL C 6.0
Product Code. 17841969 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage Unit Size
17841969 100/115/220/230 V Each
1 options
Product Code. 17841969 Supplier LNI Swissgas Supplier No. 6920.25.360
  • ONLINE EXCLUSIVE PRICE
    10685.00 €
    9265.85 € / Pack of 1
    Save 1419.15 €
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Zero air generator for HC/carbon dioxide/moisture removal with integrated compressor.
  • Zero air generator 6.0 L/min HC/carbon dioxide/moisture removal with integrated compressor.
Specifications
Voltage 100/115/220/230 V
Type Zero Air Gas Generator