missing translation for 'onlineSavingsMsg'
Learn More

Zalcitabine, TRC

Product Code. 30453068
Change view
Click to view available options
Quantity:
10 mg
25 mg
1 g
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453068 1 g -
30449288 25 mg 25mg
30451009 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453068 Supplier Toronto Research Chemicals Supplier No. TRCZ1400001G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7481-89-2
Molecular Formula C9 H13 N3 O3
Molecular Weight (g/mol) 211.22
Synonym Cytidine, 2',3'-dideoxy-, 2',3'-Dideoxycytidine, D 2C, Dideoxycytidine, Hivid, NSC 606170, Ro 24-2027/000, Zalcitabine, ddC
IUPAC Name 4-amino-1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@H]2CC[C@@H](CO)O2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H13N3O3/c10-7-3-4-12(9(14)11-7)8-2-1-6(5-13)15-8/h3-4,6,8,13H,1-2,5H2,(H2,10,11,14)/t6-,8+/m0/s1
Formula Weight 211.0957
Chemical Name or Material Zalcitabine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.