missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Zalcitabine

Product Code. 16433337
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16433337 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16433337 Supplier MedChemExpress Supplier No. HY1739250mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Zalcitabine is a potent nucleoside analogue reverse transcriptase inhibitor used in the treatment of HIV infection.

Specifications

Molecular Weight (g/mol) 211.22 g/mol
Product Type Reference compound
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 98.0%
Quantity 50 mg
Solubility Information DMSO : 62.5 mg/mL (295.90 mM∣Need ultrasonic)
For Use With (Application) COVID-19-anti-virus
CAS 7481-89-2
Synonym 2',3'-Dideoxycytidine∣ ddC∣ Dideoxycytidine
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.