Learn More
Emma Safety Footwear Zandvoort XD Safety Shoes
Product Code. 15938751
Click to view available options
Size:
39
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15938751
Brand: Emma Safety Footwear MM54886645
Only null left
Add to basket
Add to basket
A mid-level boot with water repellant oiled Nubuck uppers, internal steel toe protection and an exceptional sticky grip even on wet and greasy surfaces. Primarily designed for Metal workers, the Zandvoort is used in outdoor occupations where a strong, safe, comfortable shoe is essential.
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Design Zandvoort XD
Material Oiled Nubuck
Size 45