Learn More
Emma Safety Footwear Zandvoort XD Safety Shoes
Product Code. 15968751
Change view
Click to view available options
Size:
39
44
45
46
47
48
Unit Size:
1 pair
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15968751 48 1 pair
15918751 39 1 pair
15928751 44 1 pair
15938751 45 1 pair
15948751 46 1 pair
15958751 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15968751 Supplier Emma Safety Footwear Supplier No. MM54886648
  • ONLINE EXCLUSIVE PRICE
    178.50 €
    111.30 € / 1 pairs
    Save 67.20 €
    37% Off
Estimated Shipment: 24-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A mid-level boot with water repellant oiled Nubuck uppers, internal steel toe protection and an exceptional sticky grip even on wet and greasy surfaces. Primarily designed for Metal workers, the Zandvoort is used in outdoor occupations where a strong, safe, comfortable shoe is essential.
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Design Zandvoort XD
Material Oiled Nubuck
Size 48