missing translation for 'onlineSavingsMsg'
Learn More

Zaprinast, 98+%

Product Code. 15435589
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435589 10 mg 10mg
15455589 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435589 Supplier Thermo Scientific Alfa Aesar Supplier No. J63326.MA
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 37762-06-4 | C13H13N5O2 | 271.28 g/mol

Phosphodiesterase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37762-06-4
Molecular Formula C13H13N5O2
Molecular Weight (g/mol) 271.28
MDL Number MFCD00214073
InChI Key REZGGXNDEMKIQB-UHFFFAOYSA-N
Synonym zaprinast, zaprinastum inn-latin, unii-gxt25d5ds0, 8-aza-2-2-propoxyphenyl-6-purinone, tocris-0947, 2-o-propoxyphenyl-8-azapurine-6-one, 2-o-propoxyphenyl-8-azapurin-6-one, gxt25d5ds0, 2-2-propoxyphenyl-8-aza-6-purinone, 5-2-propoxyphenyl-1h-1,2,3 triazolo 4,5-d pyrimidin-7-ol
PubChem CID 5722
IUPAC Name 5-(2-propoxyphenyl)-2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one
SMILES CCCOC1=CC=CC=C1C2=NC(=O)C3=NNNC3=N2

Specifications

Packaging Glass bottle
Quantity 10 mg
Formula Weight 271.28
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Zaprinast

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.