missing translation for 'onlineSavingsMsg'
Learn More

Zaprinast, TRC

Product Code. 30457339
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457339 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457339 Supplier Toronto Research Chemicals Supplier No. TRCZ1500001G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zaprinast
CAS 37762-06-4
Purity Grade Notes HPLC
Molecular Formula C13 H13 N5 O2
Quantity 1 g
InChI Formula InChI=1S/C13H13N5O2/c1-2-7-20-9-6-4-3-5-8(9)11-14-12-10(13(19)15-11)16-18-17-12/h3-6H,2,7H2,1H3,(H2,14,15,16,17,18,19)
SMILES CCCOc1ccccc1C2=NC(=O)c3[nH]nnc3N2
IUPAC Name 5-(2-propoxyphenyl)-1,4-dihydrotriazolo[4,5-d]pyrimidin-7-one
Molecular Weight (g/mol) 271.27
Formula Weight 271.1069
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.