missing translation for 'onlineSavingsMsg'
Learn More

Zardaverine, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18015462
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015462 10 mg 10mg
30002152 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18015462 Supplier Tocris Bioscience Supplier No. 1046/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

PDE3/4 inhibitor

Phosphodiesterase inhibitor, selective for PDE3 and 4 (IC50 values are 0.5 and 0.8 μM respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101975-10-4
Molecular Formula C12H10F2N2O3
Molecular Weight (g/mol) 268.22
InChI Key HJMQDJPMQIHLPB-UHFFFAOYSA-N
Synonym zardaverine, zardaverina, zardaverinum, zardaverine inn, zardaverinum inn-latin, unii-tq358gwh6y, zardaverina inn-spanish, 6-4-difluoromethoxy-3-methoxy-phenyl-2h-pyridazin-3-one, 6-4-difluoromethoxy-3-methoxyphenyl-3 2h-pyridazinone, tq358gwh6y
PubChem CID 5723
ChEBI CHEBI:46548
IUPAC Name 3-[4-(difluoromethoxy)-3-methoxyphenyl]-1H-pyridazin-6-one
SMILES COC1=C(C=CC(=C1)C2=NNC(=O)C=C2)OC(F)F

Specifications

Quantity 10 mg
Formula Weight 268.22
Percent Purity >98%
Chemical Name or Material Zardaverine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.