missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ZD 7155 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ZD 7155 hydrochloride is a potent and selective competitive antagonist for the angiotensin II type 1 (AT1) receptor. Displaces [125I]-angiotensin II binding with an IC50 value of 3.8 nM in guinea pig adrenal gland membranes. Orally active, and is more potent and longer-acting than the prototype AT1 antagonist, losartan.
Specifications
Specifications
| Chemical Name or Material | 5,7-Diethyl-3,4-dihydro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1,6-naphthyridin-2(1H)-one hydrochloride |
| CAS | 146709-78-6 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Solubility | Soluble to 25mM in ethanol |
| Molecular Formula | C26H26N6O.HCl |
| Formula Weight | Observed MW: 474.99 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction