missing translation for 'onlineSavingsMsg'
Learn More

Melford Zeatin Riboside

Product Code. p-200087120
Click to view available options
Quantity:
25 mg
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 30018973

Brand: Melford Z114021.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Naturally occurring cytokinin riboside.

Naturally occurring cytokinin riboside. Main effects in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Chromatographically purified and recrystallized. Plant cell culture tested. Mixed isomers: trans-isomer 97%, cis-isomer >2%.

Specifications

CAS 6025-53-2
Quantity 1 g
Molecular Formula C15H21N5O5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.