missing translation for 'onlineSavingsMsg'
Learn More

Zeaxanthin, 65%, TRC

Product Code. 17987154
Change view
Click to view available options
Quantity:
1 mg
10 mg
250 mg
Unit Size:
10mg
1mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17987154 1 mg 1mg
17938794 250 mg 250mg
17948114 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17987154 Supplier Toronto Research Chemicals Supplier No. TRCZ2750001MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-68-3
Molecular Formula C40 H56 O2
Molecular Weight (g/mol) 568.87
Synonym Zeaxanthin (7CI), β-Carotene-3,3'-diol, (3R,3'R)-all-trans- (6CI,8CI), (3R,3'R)-β,β-Carotene-3,3'-diol, (3R,3'R)-Zeaxanthin, Anchovyxanthin, Anchovyxanthin, all-trans-, E 161H, Luteinofta, Optisharp, Xanthophyll 3, ZeaGold, Zeaxanthin, all-trans-, Zeaxanthol, all-E-Zeaxanthin, all-trans-3R,3'R-Zeaxanthin, all-trans-Anchovyxanthin, all-trans-Zeaxanthin
IUPAC Name (1R)-4-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-18-[(4R)-4-hydroxy-2,6,6-trimethylcyclohexen-1-yl]-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaenyl]-3,5,5-trimethylcyclohex-3-en-1-ol
SMILES C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C[C@@H](O)CC1(C)C)\C=C\C=C(/C)\C=C\C2=C(C)C[C@@H](O)CC2(C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C40H56O2/c1-29(17-13-19-31(3)21-23-37-33(5)25-35(41)27-39(37,7)8)15-11-12-16-30(2)18-14-20-32(4)22-24-38-34(6)26-36(42)28-40(38,9)10/h11-24,35-36,41-42H,25-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,29-15+,30-16+,31-19+,32-20+/t35-,36-/m1/s1
Formula Weight 568.428
Chemical Name or Material Zeaxanthin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.