missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Zeba™ Spin Desalting Columns and Plates, 40K MWCO, 75 μL–10 mL

Product Code. 17866648 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Columns
2 plates
25 columns
4 plates
5 columns
50 columns
Sample Volume (Metric):
1000 to 4000 μL
20 to 100 μL
300 to 800 μL
4 to 10 mL
5 to 14 μL
50 to 150 μL
500 to 2000 μL
Unit Size:
Pack of 10
Pack of 2
Pack of 25
Pack of 4
Pack of 5
Pack of 50
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Sample Volume (Metric) Unit Size
17866648 5 columns 300 to 800 μL Pack of 5
30245278 5 columns 4 to 10 mL Pack of 5
30245333 10 Columns 4 to 10 mL Pack of 10
17846548 25 columns 5 to 14 μL Pack of 25
17953321 50 columns 5 to 14 μL Pack of 50
17866468 25 columns 50 to 150 μL Pack of 25
17912981 50 columns 50 to 150 μL Pack of 50
17986700 25 columns 300 to 800 μL Pack of 25
17806518 5 columns 500 to 2000 μL Pack of 5
17927102 25 columns 500 to 2000 μL Pack of 25
17844688 5 columns 1000 to 4000 μL Pack of 5
18705495 25 columns 1000 to 4000 μL Pack of 25
17876648 2 plates 20 to 100 μL Pack of 2
17816518 4 plates 20 to 100 μL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17866648 Leverandør Thermo Scientific™ Leverandørnr. A57761
Black Friday Starts Here
Use promo code 27984
Se alle kampagner
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

The new and enhanced Zeba Spin and Micro Spin Desalting columns and plates are designed for efficient desalting and buffer exchanging proteins with a molecular weight >40 kDa and removing small molecules less than 2 kDa. They provide exceptional desalting and protein recovery performance.

Thermo Scientific Zeba spin and micro spin desalting columns and plates are designed for efficient desalting and buffer exchange of protein samples. They contain a proprietary size-exclusion chromatography resin with a robust bead structure that withstands centrifugal force and helps produce better flow characteristics and increased efficiency in removing low molecular weight protein contaminants from samples. The Zeba resin performs well even with significantly diluted protein samples and eliminates over 95% of contaminants without causing protein loss.

Separating macromolecules, such as proteins from small contaminants, is often done through routine methods including gel filtration chromatography and buffer exchange. These methods are commonly used during protein purification workflows to remove small contaminants like salts, excess labels, and cross-linking reagents from samples before analysis.

The new and enhanced Thermo Scientific Zeba Spin Desalting columns and plates are specifically designed for efficient desalting and buffer exchanging proteins with a molecular weight >40 kDa and removing small molecules less than 2 kDa. They contain a proprietary and enhanced size-exclusion chromatography resin with a more robust bead structure, making them ideal for handling centrifugal force pressure. The strengthened bead structure results in better flow characteristics and increased efficiency in removing low MW protein contaminants from samples. The Zeba resin performs exceptionally well in purifying protein samples, even when they are significantly diluted. It eliminates over 95% of contaminants without causing protein loss.

Zeba desalting columns are the ideal solution for quick and straightforward desalting and buffer exchanging of protein samples. They are offered in variousformats, such as pre-dispensed spin columns (75 μL, 0.5 mL, 2 mL, 5 mL, 10 mL), 96-well filter plates, and chromatography cartridges. These options make it simple to handle sample volumes between 2 μL and 4 mL.

Zeba Spin Desalting 96-well Plates provide trouble-free high throughput desalting and buffer exchange for sample volumes ranging from 20 to 100 μL. They require no resin dispensing or hydration, and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules. One plate can process 96 small sample volumes (20 to 100 μL) in five minutes.

Features and Benefits
• High-performing resin—exceptional protein recovery at lower protein concentrations
• Fast and Efficient—eliminate sample dilution, fraction collection, and the need for expensive chromatography system that take time to set up and can only process one sample at a time
• Easy to use—no cumbersome column preparation or equilibration
• Flexible formats—available in spin columns and 96-well spin plates for a range of sample volumes (2 μL to 4 mL)

TRUSTED_SUSTAINABILITY

Tekniske data

Product Type Spin Desalting Column
Format Spin Column
Purification Target Buffer Exchange, Protein
Column Type Size-exclusion, Proprietary Resin
MWCO 40 kDa
Product Line Zeba
Disposable Single-use
Quantity 5 columns
Volume (Metric) 2 mL
Sample Volume (Metric) 300 to 800 μL
Can Zeba Desalting columns, plates, and cartridges be autoclaved?

Yes, Zeba Desalting columns, plates, and cartridges can be autoclaved at 115 degrees C for 30 min.

What reagents and conditions are Zeba Desalting columns, plates, and cartridges compatible with?

Zeba Desalting columns, plates, and cartridges are compatible with a range of reagents and conditions. They have been confirmed to be compatible with the following:
- pH range of 3-10
- Mild oxidants/reductants
- Chaotropes (stable in 8 M urea and 6 M guanidine HCl)
- Salts
- Alcohols up to 20%
- Organic solvents such as dimethyl sulfoxide (DMSO) and dimethylformamide (DMF) (we recommend using a step-gradient of increasing concentration of the organic solvent during column equilibration)

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.