missing translation for 'onlineSavingsMsg'
Learn More

Zebularine, MedChemExpress

Product Code. 16491837
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491837 50 mg 50mg
16425678 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491837 Supplier Medchem Express Supplier No. HY13420
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Zebularine (NSC309132; 4-Deoxyuridine) is a DNA methyltransferase inhibitor. Zebularine also inhibits cytidine deaminase with a Ki of 0.95 μM.

Zebularine (NSC309132

Specifications

Chemical Name or Material Zebularine
CAS 3690-10-6
Color White
Purity Grade Notes Research
Molecular Formula C9H12N2O5
Quantity 50 mg
Synonym NSC309132 4-Deoxyuridine
Solubility Information DMSO : ≥ 100 mg/mL (438.21 mM) ∣H2O : 50 mg/mL (219.11 mM; Need ultrasonic)
SMILES O=C1N=CC=CN1[C@H]2[C@@H]([C@@H]([C@@H](CO)O2)O)O
Molecular Weight (g/mol) 228.2
Formula Weight 228.2
Percent Purity 97.57%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.