Learn More
Thermo Scientific™ Zeiss 63X Objective, LD Plan-Neofluar (0.75 NA)
Expand the functionality of your Thermo Scientific™ high-content instrument
Brand:  Thermo Scientific™ N010251
Additional Details : Weight : 0.37500kg
Product Code. 17256583
  • 10350.00 € / Each
Estimated Shipment: 14-05-2024
to see stock.
Description
Description
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
Specifications
Specifications
Objective Lens
Zeiss 63X Objective, LD Plan-Neofluar (0.75 NA)
Dry
Zeiss
ArrayScan HCA Readers
63X (0.75NA)