missing translation for 'onlineSavingsMsg'
Learn More

TerraCycle™ Zero Waste Box™ Recycling Solution For Coffee Pods and Capsules, United Kingdom

Product Code. 32188512
Change view
Click to view available options
Size:
Small
Medium
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
32188512 Medium Each
32188507 Small Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32188512 Supplier TerraCycle™ Supplier No. CS8MUK
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Support sustainability goals with an easy-to-use, all-inclusive service that helps reduce incinerated waste.

  • Capacity:
    • Small box capacity: 600 capsules
    • Medium box capacity: 1600 capsules
  • Do not send in:
    • Compostable capsules, biodegradable plastics, liquids, coffee or tea accessories, electronics, loose coffee grounds, coffee machine filters and hazardous waste
    • Each box is supplied with a pre-affixed return shipping label
  • How it works:
    • Order: Select a Zero Waste Box™ and place an order
    • Collect: Collect the accepted items to fill the box
    • Ship: Once full, use the pre-paid shipping label (already fastened to the box) to arrange for your waste to be collected for recycling
    • Reorder: Order your next Zero Waste Box™ to continue to recycle.
  • Recycling Process:
    • Everything collected is sorted and processed into raw materials that can be reused instead of being sent to landfill or incineration
    • Cardboard boxes are made from 100% recycled material

Specifications

Capacity (Metric) 78 L
Certifications/Compliance FSC
Capacity (English) 20.61 gal.
Color Brown, Green
Dimensions (L x W x H) 30.5 x 30.5 x 84 cm
Disposable Single-use
Height (Metric) 84 cm
Latex Free Yes
Length (Metric) 30.5 cm
Size Medium
Sterility Non-sterile
Width (Metric) 30.5 cm
Autoclavable Not Autoclavable
Includes Return Shipping Label
For Use With (Application) Recycling Waste
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.