missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ZFYVE19 Recombinant Protein

Product Code. 16116193
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16116193 10 μg 10µg
16126193 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16116193 Supplier Abnova™ Supplier No. H00084936P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH15738.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 84936
Molecular Weight (g/mol) 69.3
Name ZFYVE19 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MESRCYGCAVKFTLFKKEYGCKNCGRAFCSGCLSFSAAVPRTGNTQQKVCKQCHEVLTRGSSANASKWSPPQNYKKRVAALEAKQKPSTSQSQGLTRQDQMIAERLARLRQENKPKLVPSQAEIEARLAALKDERQGSIPSTQEMEARLAALQGRVLPSQTPQPAHHTPDTRTQAQQTQDLLTQLAAEVAIDESWKGGGPAASLQNDLNQGGPGSTNSKRQANWSLEEEKSRLLAEAALELREENTRQERILALAKRLAMLRGQDPERVTLQDYRLPDSDDDEDEETAIQRVLQQLTEEAALDEASGFNIPAEQASRPWTQPRGAEPEAQDVDPRPEAEEEELPWCCICNEDATLRCAGCDGDLFCARCFREGHDAFELKEHQTSAYSPPRAGQEH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FLJ14840/MPFYVE
Common Name ZFYVE19
Gene Symbol ZFYVE19
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.