missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Zileuton

Product Code. 18095302
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095302 50 mg 50mg
18083982 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095302 Supplier R&D Systems™ Supplier No. 3308/50
  • ONLINE EXCLUSIVE PRICE
    319.21 €
    287.70 € / 50mg
    Save 31.51 €
    9% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. 5-LOX inhibitor; orally bioavailable

Orally active 5-lipoxygenase (5-LOX) inhibitor that inhibits LTB4 synthesis (IC50 values are 0.56, 2.3 and 2.6 μM in dog, rat and human blood respectively). Inhibits antigen-induced contraction of tracheal strips in vitro (IC50 = 6 μM) and exhibits antiasthmatic activity in vivo. Also weakly inhibits CYP1A2 (Ki = 66 - 98 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111406-87-2
Molecular Formula C11H12N2O2S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 236.29
Purity >99%
Molecular Weight (g/mol) 236.29
Chemical Name or Material Zileuton
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.