missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Zileuton

Product Code. 16671832 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16671832 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16671832 Supplier Thermo Scientific Chemicals Supplier No. 463300010
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111406-87-2 | C11H12N2O2S | 236.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 111406-87-2
Molecular Formula C11H12N2O2S
MDL Number MFCD00866097
Quantity 1 g
InChI Key MWLSOWXNZPKENC-UHFFFAOYNA-N
SMILES CC(N(O)C(N)=O)C1=CC2=CC=CC=C2S1
IUPAC Name 1-[1-(1-benzothiophen-2-yl)ethyl]-1-hydroxyurea
Molecular Weight (g/mol) 236.29
Formula Weight 236.29
Chemical Name or Material Zileuton

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.