missing translation for 'onlineSavingsMsg'
Learn More

Zileuton, TRC

Product Code. 30457179
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457179 5 mg 5mg
30454809 10 mg 10mg
30450845 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457179 Supplier Toronto Research Chemicals Supplier No. TRCZ4200005MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111406-87-2
Molecular Formula C11 H12 N2 O2 S
Molecular Weight (g/mol) 236.29
Synonym Zileuton, Urea, N-(1-benzo[b]thien-2-ylethyl)-N-hydroxy- (9CI, ACI), N-(1-Benzo[b]thien-2-ylethyl)-N-hydroxyurea (ACI), 1-[1-(1-Benzothiophen-2-yl)ethyl]-1-hydroxyurea, A 64077, Abbott 64077, Griluto-CR, Leutrol, Zyflo
IUPAC Name 1-[1-(1-benzothiophen-2-yl)ethyl]-1-hydroxyurea
SMILES CC(N(O)C(=O)N)c1cc2ccccc2s1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H12N2O2S/c1-7(13(15)11(12)14)10-6-8-4-2-3-5-9(8)16-10/h2-7,15H,1H3,(H2,12,14)
Formula Weight 236.0619
Chemical Name or Material Zileuton
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.