missing translation for 'onlineSavingsMsg'
Learn More

Zinc, 98+%, dust (stable acc. to UN classification class 4)

Product Code. 10139232 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
10 kg
25 kg
Unit Size:
100g
10kg
1kg
2.5kg
25kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10139232 100 g 100g
10276490 1 kg 1kg
10645082 2.5 kg 2.5kg
10658124 10 kg 10kg
15224674 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10139232 Supplier Thermo Scientific Acros Supplier No. 198341000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-66-6 | Zn | 65.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6 , 1314-13-2
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Melting Point 419.0°C
Boiling Point 906.0°C
Density 7.1400g/mL
Quantity 100 g
Packaging Plastic bottle
Assay Percent Range 98+%
Form Very Fine Powder
Linear Formula Zn
Fieser 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406
Formula Weight 65.38
Percent Purity ≥98%
Chemical Name or Material Zinc
Show More Show Less
compliance-icons
Product Identifier
  • Zinc dust (stabilized)
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.