missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate, anhydrous, 99.9+%

Product Code. p-7032322
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
15476587 10 g 10g
15486587 50 g 50g
15496587 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 15476587

Brand: Thermo Scientific Alfa Aesar H33734.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 557-34-6 | C4H6O4Zn | 183.468 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Zinc acetate anhydrous is used in the synthesis of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis. It is utilized in the ultrasonic preparation of zinc sulfide nanoparticles coated on silica particles. It is administered orally or parenterally as a nutritional supplement. It finds an application in the field of industries such as wood preservation, manufacturing other zinc salts, polymers, manufacture of ethylene acetate, as a dye mordant, and analytical reagent. It also acts as a plating inhibitor on primary water piping.

Solubility
Soluble in water, alcohol, dilute mineral acids and alkalies.

Notes
Incompatible with zinc salts, alkalies and their carbonates, oxalates, phosphates, and sulfides. Hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-34-6
Molecular Formula C4H6O4Zn
Molecular Weight (g/mol) 183.468
MDL Number MFCD00012454
InChI Key DJWUNCQRNNEAKC-UHFFFAOYSA-L
Synonym zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4
PubChem CID 11192
ChEBI CHEBI:62984
IUPAC Name zinc;diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Zn+2]

Specifications

Melting Point 200°C (decomposition)
Linear Formula Zn(OOCCH3)2
Quantity 10 g
UN Number UN3077
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,dilute mineral acids and alkalies.
Formula Weight 183.47
Percent Purity ≥99.9%
Chemical Name or Material Zinc acetate, anhydrous
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc acetate, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Zinc acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.