missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, 97+%

Product Code. 11419723
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
Unit Size:
1000g
100g
2.5kg
5000g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419723 5 kg 5000g
11443890 100 g 100g
11453890 500 g 500g
11409723 1 kg 1000g
11379216 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11419723 Supplier Thermo Scientific Alfa Aesar Supplier No. A12909.0I
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5970-45-6 | C4H10O6Zn | 219.498 g/mol

Zinc acetate dihydrate is used in the preparation of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis and zinc sulfide nanoparticles coated on silica particles. It is used in dietary and medical applications. It is also used as lozenges for treating the common cold. It is used as a seeding material to grow zinc oxide nanorods on the microfibers of polyethylene terephthalate fabric. In addition, it finds application as wood preservation, production of other zinc salts, ethylene acetate manufacture, and an analytical reagent in various industries. As a plating inhibitor, it is used in nuclear power plant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and alcohol.

Notes
Incompatible with zinc salts, alkalies-their carbonates, oxalates, phosphates and sulfides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate

Specifications

Melting Point 237°C
Density 1.735
pH 6 to 7
Odor Weak, Characteristic
Quantity 5 kg
UN Number UN3077
Beilstein 3732513
Merck Index 14,10128
Solubility Information Soluble aq. soln. and alcohol.
SMILES O.O.[Zn++].CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 219.50
Formula Weight 219.50 (183.47 Anhydrous)
Percent Purity ≥97%
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.