missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Dihydrate, ACS Reagent, 98 to 101%, Honeywell Fluka™

Product Code. 15635290 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Drum with Inliner
Poly Bottle
Unit Size:
12kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15635290 500 g Poly Bottle 500g
15625290 2.5 kg Poly Bottle 2.5kg
15615290 12 kg Drum with Inliner 12kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15635290 Supplier Honeywell-Fluka Supplier No. 383058500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Melting Point ∼100°C
Color Gray
pH 6 to 8 (at 20°C)
Quantity 500 g
Linear Formula Zn(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3732513
Solubility Information In water 311g/L at 20°C
Formula Weight 219.51g/mol
Percent Purity 98.0 to 101.0%
Odor Vinegar-like
Packaging Poly Bottle
Appearance Crystalline
Density 1.750g/cm3 at 20°C
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.