missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, ACS Reagent, ≥98%, Solstice

Product Code. 16615853
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16615853 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16615853 Supplier Fluka Supplier No. 38305825KG
  • ONLINE EXCLUSIVE PRICE
    2955.00 €
    2668.42 € / 25kg
    Save 286.58 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 100°C
CAS 5970-45-6
Color Gray
pH 6 to 8
Physical Form Crystalline
Quantity 25 kg
Molecular Formula C4H10O6Zn
UN Number UN3077
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILES O.O.[Zn++].CC([O-])=O.CC([O-])=O
IUPAC Name zinc(2+) diacetate dihydrate
Molecular Weight (g/mol) 219.50
Percent Purity ≥98%
Grade ACS
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.