missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, Puriss., meets analytical spec. of DAB, E 650, 99-102%, Solstice
BIOPHARMA

Product Code. 15622730
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15622730 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15622730 Supplier Honeywell-Fluka Supplier No. 250441KG
  • ONLINE EXCLUSIVE PRICE
    119.00 €
    106.78 € / 1kg
    Save 12.22 €
    10% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of DAB, E 650, 99-102%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 5970-45-6
Physical Form Powder
Quantity 1 kg
Molecular Formula C4H10O6Zn
Linear Formula Zn(CH3COO)2 · 2H2O
MDL Number MFCD00066961
UN Number 3077
Beilstein 3732513
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
IUPAC Name zinc;diacetate;dihydrate
Molecular Weight (g/mol) 219.498
PubChem CID 2724192
Formula Weight 219.51g/mol
Percent Purity 99 to 102%
Grade puriss.
Chemical Name or Material Zinc Acetate Dihydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc acetate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Zinc acetate-2-hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.