missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Dihydrate, Reagent Grade, Honeywell™

Product Code. 15618210 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15618210 500 g Poly Bottle 500g
15608210 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15618210 Supplier Honeywell Research Chemicals Supplier No. Z0625500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Melting Point ∼100°C
Color Gray
pH 6.0 to 8.0 (at 20°C)
Quantity 500 g
Linear Formula Zn(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3732513
Formula Weight 219.51g/mol
Odor Vinegar-like
Packaging Poly Bottle
Appearance Crystalline
Density 1.750g/cu.cm at 20°C
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.