missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, Technical, ≥99%, Honeywell

Product Code. 15642730 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Paper / Poly Bag
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15642730 25 kg Paper / Poly Bag 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15642730 Supplier Honeywell Research Chemicals Supplier No. 2505625KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 5970-45-6
Quantity 25 kg
Molecular Formula C4H10O6Zn
Linear Formula Zn(CH3COO)2 · 2H2O
MDL Number MFCD00066961
UN Number UN3077
Beilstein 3732513
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
IUPAC Name zinc;diacetate;dihydrate
Molecular Weight (g/mol) 219.498
PubChem CID 2724192
Formula Weight 219.51g/mol
Percent Purity >99%
Grade Technical
Packaging Paper / Poly Bag
Chemical Name or Material Zinc acetate dihydrate, Technical, ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.