missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Dihydrate, TRC

Product Code. 30456171
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456171 1 g 1g
30454703 5 g 5g
30452337 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456171 Supplier Toronto Research Chemicals Supplier No. TRCZ7015631G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5970-45-6
Molecular Formula 2 C2 H3 O2 . Zn . 2 H2 O
Molecular Weight (g/mol) 219.53
Synonym Acetic acid, zinc salt, hydrate (2:1:2), Acetic acid, zinc salt, dihydrate (8CI,9CI), Wilzin, Zinc acetate dihydrate, Zinc diacetate dihydrate
IUPAC Name zinc;diacetate;dihydrate
SMILES O.O.[Zn+2].CC(=O)[O-].CC(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C2H4O2.2H2O.Zn/c2*1-2(3)4;;;/h2*1H3,(H,3,4);2*1H2;/q;;;;+2/p-2
Formula Weight 217.9769
Chemical Name or Material Zinc Acetate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.