missing translation for 'onlineSavingsMsg'
Learn More

Zinc bromide, 98%, extra pure, anhydrous

Product Code. 11994201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11994201 2.5 kg Glass bottle 2.5kg
10503761 100 g Glass bottle 100g
10246450 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11994201 Supplier Thermo Scientific Acros Supplier No. 202100025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7699-45-8 | Br2Zn | 225.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.19
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
Synonym zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
PubChem CID 10421208
IUPAC Name zinc;dibromide
SMILES [Zn+2].[Br-].[Br-]

Specifications

Boiling Point 697.0°C
Melting Point 394.0°C
Color White
Physical Form Chunks and/or Powder
Quantity 2.5 kg
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula ZnBr2
Fieser 02,463; 05,762; 08,535; 09,520; 10,461; 11,600; 13,349; 15,368; 16,389
Merck Index 15,10328
Solubility Information Solubility in water: 4470g/L (20°C). Other solubilities: soluble in ether, solutions of alkali hydroxides, 1g/0.5mL 90% alcohol
Formula Weight 225.19
Percent Purity 98%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 4.22
Density 4.2200g/mL
Chemical Name or Material Zinc bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.