missing translation for 'onlineSavingsMsg'
Learn More

Zinc bromide, 99.999%, (trace metal basis), extra pure

Product Code. 10751871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10751871 2.5 g Glass bottle 2.5g
10052630 10 g Glass bottle 10g
10484335 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10751871 Supplier Thermo Scientific Acros Supplier No. 212770025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7699-45-8 | Br2Zn | 225.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.19
MDL Number MFCD00011294
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
Synonym zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
PubChem CID 10421208
IUPAC Name zinc;dibromide
SMILES [Zn+2].[Br-].[Br-]

Specifications

Boiling Point 697.0°C
Melting Point 394.0°C
Color White
Physical Form Crystalline Powder
Quantity 2.5 g
Assay Percent Range 99.999%
Linear Formula ZnBr2
Fieser 02,463; 05,762; 08,535; 09,520; 10,461; 11,600; 13,349; 15,368; 16,389
Merck Index 15, 10328
Solubility Information Solubility in water: 4470g/L (20°C). Other solubilities: soluble in ether, solutions of alkali hydroxides, 1g/0.5mL 90% alcohol
Formula Weight 225.19
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.22
Density 4.2200g/mL
Chemical Name or Material Zinc bromide, 99.999%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.