missing translation for 'onlineSavingsMsg'
Learn More

Zinc Bromide puriss. anhydrous, ≥98 %, Solstice

Product Code. 16675723 Shop All Honeywell Fluka Products
Change view
Click to view available options
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16675723 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16675723 Supplier Honeywell Fluka Supplier No. 02128S2.5KG
  • ONLINE EXCLUSIVE PRICE
    477.00 €
    464.22 € / 2.5kg
    Save 12.78 €
    2% Off
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Currently Unavailable: We have no information when this product will be available.
This item is not returnable. View return policy

Specifications

Boiling Point 697°C (1,013 hPa)
Melting Point 394°C
CAS 7699-45-8
Color Colorless
CAS Max % 1
Quantity 2.5 kg
Molecular Formula Br2Zn
Linear Formula ZnBr2
Synonym Zinc dibromide
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES [Zn++].[Br-].[Br-]
IUPAC Name zinc(2+) dibromide
Molecular Weight (g/mol) 225.19
Grade Synthesis
Density 4.22 g/cm3 (20°C)
Chemical Name or Material Zinc bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.