missing translation for 'onlineSavingsMsg'
Learn More

Zinc Carbonate Basic, purum p.a., ≥58% Zn basis (KT), Honeywell™

Product Code. 15617580 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15617580

Brand: Honeywell Research Chemicals 96466250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥58% Zn basis (KT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5263-02-5
Molecular Formula C2H6O12Zn5
Molecular Weight (g/mol) 548.96
MDL Number MFCD00011519
InChI Key UOURRHZRLGCVDA-UHFFFAOYSA-D
IUPAC Name pentazinc(2+) hexahydroxide dicarbonate
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
pH 9.5 (at 50g/L 20°C)
Quantity 250 g
Linear Formula [ZnCO3]2 · [Zn(OH)2]3
UN Number NONH for all modes of transport
Percent Purity ≥58% Zn basis (KT)
Packaging Poly Bottle
Appearance Powder
Density 3.5g/cm3
Chemical Name or Material Zinc carbonate basic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.