missing translation for 'onlineSavingsMsg'
Learn More

Zinc carbonate hydroxide, 58%, extra pure, powder, (53% Zn min.)

Product Code. 10022830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10022830 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10022830 Supplier Thermo Scientific Acros Supplier No. 222650010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5263-02-5 | C2H6O12Zn5 | 548.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5263-02-5
Color White
Physical Form Fine Crystalline Powder
Quantity 1 kg
Assay Percent Range 53% min. (Zn) (Complexometry)
Molecular Formula C2H6O12Zn5
Linear Formula 3Zn(OH)2·2ZnCO3
MDL Number MFCD00011519
Merck Index 13, 10184
Solubility Information Solubility in water: practically insoluble
InChI Key UOURRHZRLGCVDA-UHFFFAOYSA-D
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O
IUPAC Name pentazinc(2+) hexahydroxide dicarbonate
Molecular Weight (g/mol) 548.96
Formula Weight 549.01
Percent Purity 58%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 4.39
Density 4.3900g/mL
Chemical Name or Material Zinc carbonate hydroxide, (53% Zn min.)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Zinc carbonate hydroxide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Zinc carbonate hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.