missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, puriss., Honeywell Fluka™
Tous les produits Honeywell Fluka Produits

Click to view available options
Quantity:
500 g
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Conditionnement:
1kg
2.5kg
25kg
500g
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécification
Specifications
| Quantity | 25 kg |
| Linear Formula | ZnCl2 |
| UN Number | 2331 |
| Formula Weight | 136.3g/mol |
| Percent Purity | 98 to 100.5% |
| Grade | puriss. |
| Vapor Pressure | 1mmHg (428°C) |
| Chemical Name or Material | Zinc Chloride, Meets analytical specification of Ph. Eur.BP, USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction