missing translation for 'onlineSavingsMsg'
Learn More

Zinc chromate

Product Code. 11483697
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483697 250 g 250g
11473697 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483697 Supplier Thermo Scientific Alfa Aesar Supplier No. A18178.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13530-65-9 | CrO4Zn | 181.37 g/mol

Zinc chromate is used in pigments, rubber, linoleum manufacturing, metal surface treatment, electroplating intermediates, research and development. It finds its application as an useful metal for preventing corrosion when applied to other metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc chromate is used in pigments, rubber, linoleum manufacturing, metal surface treatment, electroplating intermediates, research and development. It finds its application as an useful metal for preventing corrosion when applied to other metals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13530-65-9
Molecular Formula CrO4Zn
Molecular Weight (g/mol) 181.37
MDL Number MFCD00036281
InChI Key NDKWCCLKSWNDBG-UHFFFAOYSA-N
Synonym zinc chromate, zinc chrome, zinc chromate c, zinc chromate o, zinc chromate t, zinc chromate z, zinc chromate am, zincro zto, basic zinc chromate, zinc chrome yellow
PubChem CID 26089
IUPAC Name zinc;dioxido(dioxo)chromium
SMILES [Zn++].[O-][Cr]([O-])(=O)=O

Specifications

Density 3.4
Quantity 250 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 181.37
Chemical Name or Material Zinc chromate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.