missing translation for 'onlineSavingsMsg'
Learn More

Zinc citrate dihydrate, Reagent Grade, Zn 31% min

Product Code. 11323417
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323417 1 kg 1kg
11313417 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323417 Supplier Thermo Scientific Alfa Aesar Supplier No. 018129.A1
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5990-32-9 | C12H14O16Zn3 | 610.368 g/mol

Zinc citrate dihydrate is used in dental care products such as toothpastes, mouthwashes and chewing gums due to its antimicrobial and anti-inflammatory effects. It used in used in food supplements, functional foods and beverages. It is used to formulated in pharmaceuticals, and foods as a zinc supplement. It used as intermediate in organic synthesis, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Color (APHA) 10, Evaporation Residue 0.002%, Hydrogen sulfide (H2S) P.T. (limit about 1.5ppm, Sulfur dioxide (SO2) P.T. (limit about 2.5ppm), H2O 0.05%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5990-32-9
Molecular Formula C12H14O16Zn3
Molecular Weight (g/mol) 610.368
MDL Number MFCD00150587
InChI Key OXAGUGIXGVHDGD-UHFFFAOYSA-H
Synonym zinc citrate dihydrate, unii-021ibv57il, acmc-20aj8r, citric acid zinc salt dihydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, zinc salt 2:3 , dihydrate, trizinc 2+ dicitrate dihydrate, zinc citrate, dihydrate, reagent, trizinc, ion zn2+ dicitrate dihydrate, zinc 2-hydroxypropane-1,2,3-tricarboxylate-water 3/2/2, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, zinc salt, hydrate 2:3:2
PubChem CID 16213234
IUPAC Name trizinc;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Zn+2].[Zn+2].[Zn+2]

Specifications

Melting Point 334°C (decomposition)
Quantity 1 kg
Solubility Information Soluble in dilute mineral acids and alkali hydroxides. Slightly soluble in water
Formula Weight 610.35 (574.32 Anhydrous)
Grade Reagent
Physical Form Powder
Chemical Name or Material Zinc citrate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.